Learn More
4-(Chloromethyl)-2-methyl-1,3-thiazole hydrochloride , Tech., Thermo Scientific™
Product Code. 10011004
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10011004 250 mg 250mg
11355916 250 mg 250mg
10470554 1 g 1g
10382095 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10011004 Supplier Thermo Scientific Maybridge Supplier No. CD09497CB
 Restricted Item
Estimated Shipment: 24-11-2026
Chemical Identifiers
CAS 77470-53-2
Molecular Formula C5H7Cl2NS
Molecular Weight (g/mol) 184.08
MDL Number MFCD00067725
InChI Key YGKDISJLDVGNOR-UHFFFAOYSA-N
Synonym 4-chloromethyl-2-methylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-methylthiazole hcl, 4-chloromethyl-2-methyl thiazole hydrochloride, 2-methyl-4-chloromethyl thiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole, chloride, 2-methyl-4-chloromethylthiazole hydrochloride, 4-chloromethyl-2-methyl-1,3-thiazole hcl
PubChem CID 2734203
IUPAC Name 4-(chloromethyl)-2-methyl-1,3-thiazole;hydrochloride
SMILES Cl.CC1=NC(CCl)=CS1
Specifications
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 184.09
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(Chloromethyl)-2-methyl-1, 3-thiazole hydrochloride