missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorocyclohexene, TRC

Product Code. 30349022
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30349022 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349022 Supplier Toronto Research Chemicals Supplier No. TRCC371315500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 930-65-4
Molecular Formula C6H9Cl
Quantity 500 mg
Synonym Cyclohexene, 4-chloro- (6CI, 7CI, 8CI, 9CI, ACI), 4-Chlorocyclohexene (ACI)
InChI Formula InChI=1S/C6H9Cl/c7-6-4-2-1-3-5-6/h1-2,6H,3-5H2
SMILES ClC1CCC=CC1
IUPAC Name 4-chlorocyclohexene
Molecular Weight (g/mol) 116.59
Formula Weight 116.0393
Chemical Name or Material 4-Chlorocyclohexene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.