missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl Methyl Sulfone, TRC

Product Code. 30367816
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367816 500 mg 500mg
30359159 1 g 1g
30367519 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367816 Supplier Toronto Research Chemicals Supplier No. TRCC650980500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5925-80-4
Molecular Formula C8 H9 Cl O2 S
Molecular Weight (g/mol) 204.67
Synonym Benzene, 1-chloro-4-[(methylsulfonyl)methyl]-, Sulfone, p-chlorobenzyl methyl (6CI,7CI,8CI), 1-Chloro-4-[(methylsulfonyl)methyl]benzene, 4-Chlorobenzyl methyl sulfone
IUPAC Name 1-chloro-4-(methylsulfonylmethyl)benzene
SMILES CS(=O)(=O)Cc1ccc(Cl)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H9ClO2S/c1-12(10,11)6-7-2-4-8(9)5-3-7/h2-5H,6H2,1H3
Formula Weight 204.0012
Chemical Name or Material 4-Chlorobenzyl methyl sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.