Learn More
4-Chlorobenzyl methyl sulfone 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30327464
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327464 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327464 Supplier Dr. Ehrenstorfer Supplier No. DREA11392950AL100
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Chlorobenzyl methyl sulfone
CAS 5925-80-4
Analyte or Component Names 4-Chlorobenzyl methyl sulfone
Shipping Condition Room Temperature
Molecular Formula C8 H9 Cl O2 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzene, 1-chloro-4-[(methylsulfonyl)methyl]-, Sulfone, p-chlorobenzyl methyl (6CI,7CI,8CI), 1-Chloro-4-[(methylsulfonyl)methyl]benzene, 4-Chlorobenzyl methyl sulfone
InChI Formula InChI=1S/C8H9ClO2S/c1-12(10,11)6-7-2-4-8(9)5-3-7/h2-5H,6H2,1H3
SMILES CS(=O)(=O)Cc1ccc(Cl)cc1
IUPAC Name 1-chloro-4-(methylsulfonylmethyl)benzene
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 204.67
Quantity 1 mL
Formula Weight 204.0012 g/mol