missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-6-methoxymethyl-2-phenylpyrimidine

Product Code. 11365629
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11365629 1 g 1g
11355629 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365629 Supplier Alfa Aesar™ Supplier No. H50530.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 325685-59-4
Molecular Formula C12H11ClN2O
Molecular Weight (g/mol) 234.68
MDL Number MFCD00794769
InChI Key GSSSCZAOXFZADM-UHFFFAOYSA-N
Synonym 4-chloro-6-methoxymethyl-2-phenylpyrimidine, 6-chloro-2-phenyl-4-pyrimidinyl methyl methyl ether, acmc-20aml2, chlorophenylpyrimidinylmethylmethylether, 6-chloro-2-phenylpyrimidin-4-yl methoxymethane, pyrimidine,4-chloro-6-methoxymethyl-2-phenyl, 6-chloro-2-phenyl-4-pyrimidinyl methylmethyl ether
PubChem CID 2763927
IUPAC Name 4-chloro-6-(methoxymethyl)-2-phenylpyrimidine
SMILES COCC1=CC(Cl)=NC(=N1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 234.69
Chemical Name or Material 4-Chloro-6-methoxymethyl-2-phenylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.