missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11476936
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476936 5 g 5g
11466936 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476936 Supplier Thermo Scientific Alfa Aesar Supplier No. B24022.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-3-(trifluoromethyl)benzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34328-46-6
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD00040722
InChI Key NIHMMULLFBKTOK-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzaldehyde, 4-chloro-3-trifluoromethyl-benzaldehyde, 2-chloro-5-formylbenzotrifluoride, benzaldehyde, 4-chloro-3-trifluoromethyl, pubchem2892, acmc-1adw8, timtec-bb sbb003468, 4-chloro-3trifluoromethyl-benzaldehyde, 3-trifluoromethyl-4-chloro-benzaldehyde, 4-chloranyl-3-trifluoromethyl benzaldehyde
PubChem CID 118642
IUPAC Name 4-chloro-3-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1C=O)C(F)(F)F)Cl

Specifications

Boiling Point 54°C (10 mmHg)
Refractive Index 1.4964
Quantity 5 g
Beilstein 2101897
Sensitivity Air Sensitive
Formula Weight 208.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.