missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrocinnamic Acid Methyl Ester, TRC

Product Code. 30361774
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361774 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361774 Supplier Toronto Research Chemicals Supplier No. TRCC59716350G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-3-nitrocinnamic Acid Methyl Ester
CAS 877065-30-0
Molecular Formula C10H8ClNO4
Quantity 50 g
InChI Formula InChI=1S/C10H8ClNO4/c1-16-10(13)5-3-7-2-4-8(11)9(6-7)12(14)15/h2-6H,1H3/b5-3+
SMILES COC(=O)\C=C\c1ccc(Cl)c(c1)[N+](=O)[O-]
IUPAC Name methyl (E)-3-(4-chloro-3-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol) 241.63
Formula Weight 241.0142
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.