missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzyl alcohol, 98%

Product Code. 11468656
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468656 25 g 25g
11458656 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468656 Supplier Thermo Scientific Alfa Aesar Supplier No. A16024.14
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 55912-20-4 | C7H6ClNO3 | 187.579 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55912-20-4
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
MDL Number MFCD00007086
InChI Key QLLRQJDSYJIXTN-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzyl alcohol, 4-chloro-3-nitrophenyl methanol, 4-chloro-3-nitrobenzylalcohol, 4-chloro-3-nitrobenzenemethanol, 3-nitro-4-chlorobenzyl alcohol, acmc-1ayo8, qllrqjdsyjixtn-uhfffaoysa, benzenemethanol,4-chloro-3-nitro, benzenemethanol, 4-chloro-3-nitro, 4-chloro-3-nitro-phenyl-methanol
PubChem CID 91933
IUPAC Name (4-chloro-3-nitrophenyl)methanol
SMILES C1=CC(=C(C=C1CO)[N+](=O)[O-])Cl

Specifications

Melting Point 62°C to 64°C
Quantity 25 g
Formula Weight 187.58
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.