Learn More
4-chloro-3-cyanophenylboronic acid, 95%, Thermo Scientific™
Product Code. p-4228499 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10573658 1 g Glass bottle 1g
10669009 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10573658 Supplier Thermo Scientific Acros Supplier No. 430420010
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 871332-95-5
Molecular Formula C7H5BClNO2
Molecular Weight (g/mol) 181.38
MDL Number MFCD08056360
InChI Key PRPLADNLEYNYFZ-UHFFFAOYSA-N
Synonym 4-chloro-3-cyanophenyl boronic acid, 5-borono-2-chlorobenzonitrile, 4-chloro-3-cyanobenzeneboronic acid, 4-chloro-3-cyano-phenyl boronic acid, 4-chloro-3-cyanophenylboronicacid, pubchem17104, acmc-209qho, ksc447q0n, 4-chloro-3-cyanobenzeneboronic acid 97+%
PubChem CID 44119831
IUPAC Name (4-chloro-3-cyanophenyl)boronic acid
SMILES OB(O)C1=CC(C#N)=C(Cl)C=C1
Specifications
CAS Min % 94.0
CAS Max % 100.0
Melting Point 300.0°C to 304.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 181.39
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-chloro-3-cyanophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-3-cyanophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.