accessibility menu, dialog, popup

UserName

CAS RN 871332-95-5

BClNHOOH

CAS RN 871332-95-5

IUPAC Name: (4-chloro-3-cyanophenyl)boronic acid
Synonyms: 4-chloro-3-cyanophenylboronic acid
Molecular Weight (g/mol): 181.38
Molecular Formula: C7H5BClNO2
InChi Key: PRPLADNLEYNYFZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(C#N)=C(Cl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)