Learn More
4-Chloro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™
Product Code. 11322996
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322996 25 g 25g
11312996 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11322996 Supplier Thermo Scientific Alfa Aesar Supplier No. L10778.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23165-53-9
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.65
MDL Number MFCD00041089
InChI Key XTYLRVPBHHRTMS-UHFFFAOYSA-N
Synonym 4-chloro-2-methylphenyl isothiocyanate, 3-chloro-6-isothiocyanatotoluene, benzene, 4-chloro-1-isothiocyanato-2-methyl, 4-chloro-2-methylphenylisothiocyanate, 4-chloro-2-methyl-phenyl isothiocyanate, 2-methyl-4-chlorophenyl isothiocyanate, 4-chloro-2-methylbenzenisothiocyanate, acmc-20amlo, chlorotrimethylsilane tmcs, 4-chloro-2-methyl isothiocyanate
PubChem CID 90021
IUPAC Name 4-chloro-1-isothiocyanato-2-methylbenzene
SMILES CC1=CC(Cl)=CC=C1N=C=S
Specifications
Melting Point 33°C to 35°C
Quantity 25 g
Odor Irritating
UN Number UN2923
Beilstein 2963683
Boiling Point 88°C to 90°C (0.4 mmHg)
Formula Weight 183.66
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylphenyl isothiocyanate