missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzoic acid, 99%, Thermo Scientific™

Product Code. 11429257
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11429257

Brand: Thermo Scientific Alfa Aesar B21332.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57479-70-6
Molecular Formula C8H6ClO3
Molecular Weight (g/mol) 185.58
MDL Number MFCD00002532
InChI Key UFEYMXHWIHFRBX-UHFFFAOYSA-M
Synonym 4-chloro-o-anisic acid, 2-methoxy-4-chlorobenzoic acid, 4-chloro-2-methoxy-benzoic acid, 4-chloro-2-methoxybenzoicacid, benzoic acid, 4-chloro-2-methoxy, 4-chloro-ortho-anisic acid, 2-methoxy-4-chloro benzoic acid, acmc-1axaj, 4-chloro-2-anisic acid, ksc494s8r
PubChem CID 93682
SMILES COC1=CC(Cl)=CC=C1C([O-])=O

Specifications

Melting Point 145°C to 148°C
Quantity 5 g
IUPAC Name 4-chloro-2-methoxybenzoate
Formula Weight 186.59
Percent Purity 99%
Chemical Name or Material 4-Chloro-2-methoxybenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.