accessibility menu, dialog, popup

UserName

CAS RN 57479-70-6

H3COClOO

CAS RN 57479-70-6

IUPAC Name: 4-chloro-2-methoxybenzoate
Synonyms: 4-chloro-2-methoxybenzoate
Molecular Weight (g/mol): 185.58
Molecular Formula: C8H6ClO3
InChi Key: UFEYMXHWIHFRBX-UHFFFAOYSA-M
SMILES: COC1=CC(Cl)=CC=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results