Learn More
4-Chloro-2-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Product Code. 11402180
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402180 10 g 10g
11492170 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11402180 Supplier Thermo Scientific Alfa Aesar Supplier No. H28872.09
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Used as intermediates for pharmaceutical and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as intermediates for pharmaceutical and agrochemicals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 160591-91-3
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD02684293
InChI Key YBNDRTRLXPEWKQ-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenylboronicacid, 2-fluoro-4-chlorophenylboronic acid, b-4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034, 4-chloro-2-fluoro-phenyl boronic acid
PubChem CID 4193595
IUPAC Name (4-chloro-2-fluorophenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)Cl)F)(O)O
Specifications
Melting Point 246°C to 250°C
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 174.37
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-fluorobenzeneboronic acid