Learn More
4-Bromophenylacetylene, 97%
Product Code. 11369635
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369635 250 mg 250mg
11359635 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11369635 Supplier Thermo Scientific Alfa Aesar Supplier No. L19082.MD
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
CAS: 766-96-1 | C8H5Br | 181.032 g/mol
4-Bromophenylacetylene, is used as the starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromophenylacetylene, is used as the starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.
Solubility
Soluble in chloroform, diethyl ether and ethanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 766-96-1
Molecular Formula C8H5Br
Molecular Weight (g/mol) 181.032
MDL Number MFCD00168822
InChI Key LTLVZQZDXQWLHU-UHFFFAOYSA-N
Synonym 4-bromophenylacetylene, benzene, 1-bromo-4-ethynyl, 1-bromo-4-ethynyl-benzene, 4-bromophenyl acetylene, 4'-bromophenyl acetylene, 4'-bromophenylacetylene, p-bromophenylacetylene, p-bromophenyl acetylene, 4-bromphenyl acetylene, 4-bromo-1-ethynylbenzene
PubChem CID 136603
IUPAC Name 1-bromo-4-ethynylbenzene
SMILES C#CC1=CC=C(C=C1)Br
Specifications
Melting Point 64°C to 67°C
Boiling Point 88°C to 90°C (16 mmHg)
Odor Odorless
Quantity 250 mg
Beilstein 1209726
Solubility Information Soluble in chloroform,diethyl ether and ethanol.
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material 4-Bromophenylacetylene