missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisoindoline hydrochloride, 97%

Product Code. p-4228449 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10575870 1 g Glass bottle 1g
10725652 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10575870 Supplier Thermo Scientific Acros Supplier No. 435870010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 923590-95-8 | C8H8BrN·ClH | 234.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923590-95-8
Molecular Formula C8H8BrN·ClH
Molecular Weight (g/mol) 234.52
InChI Key FQHLHVFOJBANKY-UHFFFAOYSA-N
Synonym 4-bromoisoindoline hydrochloride, 4-bromo-isoindoline hcl, 4-bromo-2,3-dihydro-1h-isoindole hydrochloride, 4-bromo-1h-isoindoline hydrochloride, 4-bromoisoindoline hcl, 1h-isoindole, 4-bromo-2,3-dihydro-, hydrochloride 1:1, pubchem14849, acmc-209y8q, 4-bromo-isoindoline hydrochloride
PubChem CID 44228644
IUPAC Name 4-bromo-2,3-dihydro-1H-isoindole;hydrochloride
SMILES C1C2=C(CN1)C(=CC=C2)Br.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 269°C to 274°C
Color White to Yellow
Elemental Analysis 39.33 to 42.61 % C 5.73 to 6.21 % N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 234.52
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoisoindoline hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromoisoindoline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.