Learn More
(4-Bromobutyl)triphenylphosphonium bromide, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 7333-63-3 |
| Molecular Formula | C22H23Br2P |
| MDL Number | MFCD00031636 |
| Quantity | 25 g |
| SMILES | BrCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-] |
| Molecular Weight (g/mol) | 478.20 |
| Percent Purity | 97% |
| Chemical Name or Material | (4-Bromobutyl)triphenylphosphonium bromide |