missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde dimethyl acetal, 97%, Thermo Scientific™

Product Code. p-7052682
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491488 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15491488 Supplier Thermo Scientific Alfa Aesar Supplier No. H55283.AC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Bromobenzaldehyde dimethyl acetal, is used to produce other chemicals, it is used to produce 1-Bromo-4-bromomethyl-benzene at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24856-58-4
Molecular Formula C9H11BrO2
Molecular Weight (g/mol) 231.089
MDL Number MFCD01320499
InChI Key TWFNGPDYMFEHOB-UHFFFAOYSA-N
Synonym 4-bromobenzaldehyde dimethyl acetal, 1-bromo-4-dimethoxymethyl benzene, acmc-20ammy, pubchem5380, 4-bromophenyl dimethoxymethane, 1-bromo-4-dimethoxymethyl-benzene, 4-bromobenzaldehyde dimethyl ketal, 4-bromo-benzaldehyde dimethyl ketal
PubChem CID 688342
IUPAC Name 1-bromo-4-(dimethoxymethyl)benzene
SMILES COC(C1=CC=C(C=C1)Br)OC

Specifications

Density 1.383g/mL
Boiling Point 254°C
Refractive Index 1.5320
Quantity 25 mL
Formula Weight 231.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromobenzaldehyde dimethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.