accessibility menu, dialog, popup

UserName

CAS RN 24856-58-4

IUPAC Name: 1-bromo-4-(dimethoxymethyl)benzene
Synonyms: 1-bromo-4-(dimethoxymethyl)benzene
Molecular Weight (g/mol): 231.09
Molecular Formula: C9H11BrO2
InChi Key: TWFNGPDYMFEHOB-UHFFFAOYSA-N
SMILES: COC(OC)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results