missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-4-chlorobutyrophenone, 98%

Product Code. 11401167
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401167 100 g 100g
11491157 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11401167 Supplier Thermo Scientific Alfa Aesar Supplier No. B24146.22
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4559-96-0 | C10H10BrClO | 261.54 g/mol

4'-Bromo-4-chlorobutyrophenone is used as a pharmaceutical & syntheses material intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromo-4-chlorobutyrophenone is used as a pharmaceutical & syntheses material intermediates.

Solubility
Insoluble in water.

Notes
Storage temperature is 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4559-96-0
Molecular Formula C10H10BrClO
Molecular Weight (g/mol) 261.54
MDL Number MFCD00001006
InChI Key WJKPUMBLABGUCQ-UHFFFAOYSA-N
Synonym 1-4-bromophenyl-4-chlorobutan-1-one, 4'-bromo-4-chlorobutyrophenone, 1-4-bromophenyl-4-chloro-1-butanone, 1-butanone, 1-4-bromophenyl-4-chloro, acmc-20amn1, 4-chloro-p-bromobutyrophenone, 4-chloro-4'-bromobutyrophenone, 4-bromo-4-chlorobutyrophenone, 4/'-bromo-4-chlorobutyrophenone, 4'-bromo-gamma-chlorobutyrophenone
PubChem CID 78317
IUPAC Name 1-(4-bromophenyl)-4-chlorobutan-1-one
SMILES ClCCCC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 35°C to 38°C
Flash Point >110°C (230°F)
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 261.55
Percent Purity 98%
Chemical Name or Material 4'-Bromo-4-chlorobutyrophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.