Learn More
4-Bromo-2-(morpholin-4-yl)benzonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-2-(morpholin-4-yl)benzonitrile |
| CAS | 1260762-06-8 |
| Molecular Formula | C11H11N2OBr |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C11H11BrN2O/c12-10-2-1-9(8-13)11(7-10)14-3-5-15-6-4-14/h1-2,7H,3-6H2 |
| SMILES | C1COCCN1C2=C(C=CC(=C2)Br)C#N |
| IUPAC Name | 4-bromo-2-morpholin-4-ylbenzonitrile |
| Molecular Weight (g/mol) | 267.12 |
| Formula Weight | 266.0055 |