missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxyaniline, 98%, Thermo Scientific Chemicals

Product Code. 15467998
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467998 1 g 1g
15477998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467998 Supplier Thermo Scientific Alfa Aesar Supplier No. H61454.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59557-91-4 | Molecular Formula: C7H8BrNO | Molecular Weight: 202.051 g/mol

4-Bromo-2-methoxyaniline is used as a reagent in the synthesis of 3,5-diamino-1,2,4-triazole ureas as potent anaplastic lymphoma kinase (ALK) inhibitors. It is also used as a reagent in the synthesis of chroman-3-amides as potent Rho kinase inhibitors. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methoxyaniline is used as a reagent in the synthesis of 3,5-diamino-1,2,4-triazole ureas as potent anaplastic lymphoma kinase (ALK) inhibitors. It is also used as a reagent in the synthesis of chroman-3-amides as potent Rho kinase inhibitors. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59557-91-4
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD01204266
InChI Key WRFYIYOXJWKONR-UHFFFAOYSA-N
Synonym 4-bromo-2-methoxy-phenylamine, 2-amino-5-bromoanisole, 4-bromo-o-anisidine, 4-bromo-2-methoxyphenylamine, 4-bromo-2-methoxy-aniline, 2-methoxy-4-bromoaniline, benzenamine, 4-bromo-2-methoxy, pubchem2258, acmc-209meb, 4-bromo-2-methoxy aniline
PubChem CID 459257
IUPAC Name 4-bromo-2-methoxyaniline
SMILES COC1=C(C=CC(=C1)Br)N

Specifications

Quantity 1 g
UN Number UN2811
Solubility Information Soluble in methanol.
Formula Weight 202.05
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.