Learn More
4-Bromo-1-methanesulfonyl-2-methylbenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-1-methanesulfonyl-2-methylbenzene |
| CAS | 99768-21-5 |
| Molecular Formula | C8H9BrO2S |
| Quantity | 500 mg |
| Synonym | 4-Bromo-2-methyl-1-(methylsulfonyl)benzene, 4-Bromo-o-tolyl Methylsulfone |
| InChI Formula | InChI=1S/C8H9BrO2S/c1-6-5-7(9)3-4-8(6)12(2,10)11/h3-5H,1-2H3 |
| SMILES | BrC1=CC=C(S(C)(=O)=O)C(C)=C1 |
| IUPAC Name | 4-bromo-2-methyl-1-(methylsulfonyl)benzene |
| Molecular Weight (g/mol) | 249.12 |
| Formula Weight | 247.95 |