Learn More
4-Benzyloxyacetophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Benzyloxyacetophenone |
| CAS | 54696-05-8 |
| Molecular Formula | C15H14O2 |
| Quantity | 2.5 g |
| Synonym | 1-[4-(Phenylmethoxy)phenyl]-ethanone, 4'-(Benzyloxy)-acetophenone, 1-(4-Benzyloxyphenyl)ethanone, 1-(4-Phenylmethoxyphenyl)ethanone, 4-Acetylphenyl Benzyl Ether, 4-Benzyloxyacetophenone, 4'-Benzyloxyacetophenone, NSC 56945, p-(Benzyloxy)acetophenone, p-(Phenylmethoxy)acetophenone |
| InChI Formula | InChI=1S/C15H14O2/c1-12(16)14-7-9-15(10-8-14)17-11-13-5-3-2-4-6-13/h2-10H,11H2,1H3 |
| SMILES | CC(=O)c1ccc(OCc2ccccc2)cc1 |
| IUPAC Name | 1-(4-phenylmethoxyphenyl)ethanone |
| Molecular Weight (g/mol) | 226.27 |
| Formula Weight | 226.0994 |