Learn More
4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione, TRC
Product Code. 30334990
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334990 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30334990 Supplier Toronto Research Chemicals Supplier No. TRCB28572510MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
CAS 327036-89-5
Molecular Formula C10 H10 N2 O2 S
Quantity 10 mg
Synonym TDZD 8 - Bio-X TM
InChI Formula InChI=1S/C10H10N2O2S/c1-11-9(13)12(10(14)15-11)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3
SMILES CN1SC(=O)N(Cc2ccccc2)C1=O
IUPAC Name 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
Molecular Weight (g/mol) 222.26
Formula Weight 222.0463