missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzoic acid, 97%

Product Code. 11435527
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435527 100 g 100g
11425527 25 g 25g
11445527 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435527 Supplier Thermo Scientific Alfa Aesar Supplier No. B23519.22
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-91-7 | C8H9NO2 | 151.165 g/mol

4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.

Solubility
Slightly soluble in water. Insoluble in ethanol, benzene and chloroform.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-91-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00010203
InChI Key QCTBMLYLENLHLA-UHFFFAOYSA-N
Synonym 4-aminomethyl benzoic acid, 4-carboxybenzylamine, pamba, styptopur, gumbix, p-aminomethylbenzoic acid, aminomethylbenzoic acid, benzoic acid, 4-aminomethyl, benzylamine-4-carboxylic acid, alpha-amino-p-toluic acid
PubChem CID 65526
IUPAC Name 4-(aminomethyl)benzoic acid
SMILES C1=CC(=CC=C1CN)C(=O)O

Specifications

Melting Point ∼290°C (decomposition)
Quantity 100 g
Beilstein 1100606
Solubility Information Slightly soluble in water. Insoluble in ethanol,benzene and chloroform.
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 4-(Aminomethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.