Learn More
4-Amino-nicotinonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-nicotinonitrile |
| CAS | 15827-84-6 |
| Molecular Formula | C6 H5 N3 |
| Quantity | 25 mg |
| Synonym | Nicotinonitrile, 4-amino- (6CI,8CI), 4-Amino-3-pyridinecarbonitrile, 3-Cyano-4-aminopyridine, 4-Amino-3-cyanopyridine, 4-Aminonicotinonitrile |
| InChI Formula | InChI=1S/C6H5N3/c7-3-5-4-9-2-1-6(5)8/h1-2,4H,(H2,8,9) |
| SMILES | Nc1ccncc1C#N |
| IUPAC Name | 4-aminopyridine-3-carbonitrile |
| Molecular Weight (g/mol) | 119.124 |
| Formula Weight | 119.048 |