Learn More
4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 152-47-6 |
| Molecular Formula | C11H12N4O3S |
| MDL Number | MFCD00437754 |
| Quantity | 25 g |
| SMILES | O=S(C1=CC=C(N)C=C1)(NC2=NC=CN=C2OC)=O |
| Molecular Weight (g/mol) | 280.30 |
| Percent Purity | 99% |
| Chemical Name or Material | 4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide |