Learn More
4-Amino-5-chloro-2-methoxybenzoic Acid, Mikromol™
Description
4-Amino-5-chloro-2-methoxybenzoic Acid
Specifications
Specifications
| Chemical Name or Material | 4-Amino-5-chloro-2-methoxybenzoic Acid |
| CAS | 7206-70-4 |
| Analyte or Component Names | 4-Amino-5-chloro-2-methoxybenzoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H8 Cl N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Amino-5-chloro-2-methoxybenzoic Acid, Metoclopramide Imp. C (EP), Metoclopramide Hydrochloride Monohydrate Impurity C, Metoclopramide Impurity C, Metoclopramide Impurity C |
| InChI Formula | InChI=1S/C8H8ClNO3/c1-13-7-3-6(10)5(9)2-4(7)8(11)12/h2-3H,10H2,1H3,(H,11,12) |
| SMILES | COc1cc(N)c(Cl)cc1C(=O)O |
| IUPAC Name | 4-amino-5-chloro-2-methoxybenzoic acid |
| Show More |