missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-chloro-2,1,3-benzothiadiazole, TRC

Product Code. 30133272
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133272 5 g 5g
30005257 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133272 Supplier Toronto Research Chemicals Supplier No. TRCA6034305G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30536-19-7
Molecular Formula C6 H4 Cl N3 S
Molecular Weight (g/mol) 185.63
Synonym 2,1,3-Benzothiadiazol-4-amine, 5-chloro-, 2,1,3-Benzothiadiazole, 4-amino-5-chloro- (6CI,7CI,8CI), 5-Chloro-2,1,3-benzothiadiazol-4-amine, 4-Amino-5-chloro-2,1,3-benzothiadiazole, 5-Chloro-4-amino-2,1,3-benzothiadiazole
IUPAC Name 5-chloro-2,1,3-benzothiadiazol-4-amine
SMILES Nc1c(Cl)ccc2nsnc12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H4ClN3S/c7-3-1-2-4-6(5(3)8)10-11-9-4/h1-2H,8H2
Formula Weight 184.9814
Percent Purity >95%
Chemical Name or Material 4-Amino-5-chloro-2,1,3-benzothiadiazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.