missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-hydroxydiphenylsulfone, TRC

Product Code. 17973585
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973585 25 mg 25mg
17913785 250 mg 250mg
17963485 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973585 Supplier Toronto Research Chemicals Supplier No. TRCA61122525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25963-47-7
Molecular Formula C12 H11 N O3 S
Molecular Weight (g/mol) 249.29
Synonym 4-[(4-Aminophenyl)sulfonyl]phenol, p-Sulfanilylphenol;
IUPAC Name 4-(4-aminophenyl)sulfonylphenol
SMILES Nc1ccc(cc1)S(=O)(=O)c2ccc(O)cc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H11NO3S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8,14H,13H2
Formula Weight 249.046
Percent Purity >95%
Chemical Name or Material 4-Amino-4'-hydroxydiphenylsulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.