Learn More
4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide Hydrochloride |
| CAS | 247584-10-7 |
| Molecular Formula | C8H15ClN4O |
| Quantity | 5 g |
| Synonym | 4-Amino-2-methyl-5-propyl-2H-pyrazole-3-carboxamide Hydrochloride |
| InChI Formula | InChI=1S/C8H14N4O.ClH/c1-3-4-5-6(9)7(8(10)13)12(2)11-5;/h3-4,9H2,1-2H3,(H2,10,13);1H |
| SMILES | Cl.CCCc1nn(C)c(C(=O)N)c1N |
| IUPAC Name | 4-amino-2-methyl-5-propylpyrazole-3-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 218.68 |
| Formula Weight | 218.0934 |