Learn More
4'-Acetyl Simvastatin, TRC
Product Code. 30138115
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138115 100 mg 100mg
30003560 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138115 Supplier Toronto Research Chemicals Supplier No. TRCA187795100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145576-25-6
Molecular Formula C27 H40 O6
Molecular Weight (g/mol) 460.6
Synonym (1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-(Acetyloxy)-6-oxotetrahydro-2H-pyran-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-Dimethylbutanoate, Simvastatin Imp. B (EP), Simvastatin Acetate Ester
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-acetyloxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
SMILES CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@H](CC(=O)O3)OC(=O)C)[C@@H]12
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C27H40O6/c1-7-27(5,6)26(30)33-23-13-16(2)12-19-9-8-17(3)22(25(19)23)11-10-20-14-21(31-18(4)28)15-24(29)32-20/h8-9,12,16-17,20-23,25H,7,10-11,13-15H2,1-6H3/t16-,17-,20+,21+,22-,23-,25-/m0/s1
Formula Weight 460.2825
Chemical Name or Material Acetyl Simvastatin