Learn More
4-Acetoxy-N-isopropyl-N-methyltryptamine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Acetoxy-N-isopropyl-N-methyltryptamine |
| CAS | 1024612-25-6 |
| Molecular Formula | C16H22N2O2 |
| Quantity | 50 mg |
| Synonym | 4-Acetate 3-[2-[Methyl(1-methylethyl)amino]ethyl]-1H-indol-4-ol |
| InChI Formula | InChI=1S/C16H22N2O2/c1-11(2)18(4)9-8-13-10-17-14-6-5-7-15(16(13)14)20-12(3)19/h5-7,10-11,17H,8-9H2,1-4H3 |
| SMILES | CC(C)N(C)CCc1c[nH]c2cccc(OC(=O)C)c12 |
| IUPAC Name | [3-[2-[methyl(propan-2-yl)amino]ethyl]-1H-indol-4-yl] acetate |
| Molecular Weight (g/mol) | 274.36 |
| Formula Weight | 274.1681 |