Learn More
4'-[[7-Methyl-5-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl]biphenyl-2-carboxylic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4'-[[7-Methyl-5-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl]biphenyl-2-carboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 4'-[(1,7'-dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazol-1'-yl)methyl]biphenyl-2-carboxylic acid |
| CAS | 1026353-20-7 |
| Analyte or Component Names | 4'-[(1,7'-dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazol-1'-yl)methyl]biphenyl-2-carboxylic acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H30 N4 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Telmisartan Imp. B (EP), Telmisartan USP Related Compound B, Telmisartan USP RC B, 4'-[(1,7'-Dimethyl-2'-propyl[2,5'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxylic acid, 4'-[(1,7'-dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazol-1'-yl)methyl]biphenyl-2-carboxylic acid, Telmisartan RC B (USP), Telmisartan Related Compound B, Telmisartan Impurity B |
| InChI Formula | InChI=1S/C33H30N4O2/c1-4-9-30-34-28-19-24(32-35-27-12-7-8-13-29(27)36(32)3)18-21(2)31(28)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19H,4,9,20H2,1-3H3,(H,38,39) |
| SMILES | CCCc1nc2cc(cc(C)c2n1Cc3ccc(cc3)c4ccccc4C(=O)O)c5nc6ccccc6n5C |
| IUPAC Name | 2-[4-[[7-methyl-5-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzoic acid |
| Show More |