Learn More
4,6-Dichloro-5-(2-methoxyphenoxy)-2,2’-bipyrimidine, TRC
Product Code. 17932035
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932035 100 mg 100mg
30138048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17932035 Supplier Toronto Research Chemicals Supplier No. TRCD434910100MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 150728-13-5
Molecular Formula C15 H10 Cl2 N4 O2
Molecular Weight (g/mol) 349.17
Synonym 2,2'-Bipyrimidine, 4,6-dichloro-5-(2-methoxyphenoxy)-, 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine, 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-dipyrimidine, 4,6-Dichloro-5-(o-methoxyphenoxy)-2-(2-pyrimidinyl)pyrimidine, (4,6-dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine), Bosentan Related Compound D
IUPAC Name 4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
SMILES COc1ccccc1Oc2c(Cl)nc(nc2Cl)c3ncccn3
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H10Cl2N4O2/c1-22-9-5-2-3-6-10(9)23-11-12(16)20-15(21-13(11)17)14-18-7-4-8-19-14/h2-8H,1H3
Formula Weight 348.0181
Chemical Name or Material 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine