Learn More
4,6-Diamino-2-chloro-5-pyrimidinecarbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4,6-Diamino-2-chloro-5-pyrimidinecarbonitrile |
| CAS | 18340-56-2 |
| Molecular Formula | C5H4ClN5 |
| Quantity | 50 mg |
| Synonym | 2-Chloro-4,6-diamino-5-cyanopyrimidine |
| InChI Formula | InChI=1S/C5H4ClN5/c6-5-10-3(8)2(1-7)4(9)11-5/h(H4,8,9,10,11) |
| SMILES | O=C1O[C@H](C2=CC=CC=C2)[C@@H](C)N1 |
| Molecular Weight (g/mol) | 169.57 |