Learn More
4,5-Dichloro-3-hydroxypyridazine, 98%, Thermo Scientific™
Product Code. 10586114 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10586114 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10586114 Supplier Thermo Scientific Acros Supplier No. 426670250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 932-22-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 204.0°C to 206.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C4H2Cl2N2O
MDL Number MFCD00051504
Quantity 25 g
Synonym 4,5-dichloro-3 2h-pyridazinone, 4,5-dichloropyridazin-3 2h-one, 3 2h-pyridazinone, 4,5-dichloro, 4,5-dichloropyridazin-3-ol, 4,5-dichloro-2h-pyridazin-3-one, 4,5-dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one, 4,5-dichloro-pyridazin-3-ol, 4,5-dichloro-3-pyridazinol
InChI Key VJWXIRQLLGYIDI-UHFFFAOYSA-N
SMILES ClC1=C(Cl)C(=O)NN=C1
IUPAC Name 4,5-dichloro-2,3-dihydropyridazin-3-one
Molecular Weight (g/mol) 164.97
PubChem CID 73247
Formula Weight 164.98
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 5-Dichloro-3-hydroxypyridazine