Learn More
4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine Dihydrochloride, TRC
Product Code. 30453337
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453337 2.5 g -
30454379 500 mg 500mg
30457292 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30453337 Supplier Toronto Research Chemicals Supplier No. TRCT8896932.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62002-31-7
Molecular Formula C6 H9 N3 . 2[HCl]
Molecular Weight (g/mol) 196.078
IUPAC Name 4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine;dihydrochloride
SMILES Cl.Cl.C1Cc2nc[nH]c2CN1
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C6H9N3.2ClH/c1-2-7-3-6-5(1)8-4-9-6;;/h4,7H,1-3H2,(H,8,9);2*1H
Formula Weight 195.033
Chemical Name or Material 4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine Dihydrochloride