Learn More
4,5,6,7-Tetrahydro-1H-indazole, 98%, Thermo Scientific™
Product Code. 11361307
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11361307 5 g 5g
11351307 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11361307 Supplier Thermo Scientific Alfa Aesar Supplier No. L01515.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2305-79-5
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00014565
InChI Key GDSQTWDUCDSZEY-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydroindazole, 4,5,6,7-tetrahydro-2h-indazole, 1h-indazole, 4,5,6,7-tetrahydro, 3,4-tetramethylenepyrazole, 4 5 6 7 tetrahydroindazole, 1h-4,5,6,7-tetrahydroindazole, acmc-1cl2g, 4,6,7-tetrahydroindazole, 4,6,7-tetrahydro-1h-indazole, 3,4-tetramethylene-1h-pyrazole
PubChem CID 75317
IUPAC Name 4,5,6,7-tetrahydro-1H-indazole
SMILES C1CCC2=C(C1)C=NN2
Specifications
Melting Point 80°C to 84°C
Boiling Point 140°C to 142°C (2 mmHg)
Quantity 5 g
Beilstein 2420
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 4,5,6,7-Tetrahydro-1H-indazole