missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Octahydro-4,7-methano-5H-inden-5-ylidene) bisphenol, 99%, Thermo Scientific™

Product Code. 10443171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10443171 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10443171 Supplier Thermo Scientific Acros Supplier No. 416351000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1943-97-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 219°C to 221°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C22H24O2
MDL Number MFCD00213567
Quantity 100 g
Synonym 5,7-methanoindan, 4,4'-octahydro-1h-4,7-methanoindene-5,5-diyl diphenol, phenol, 4,4'-octahydro-4,7-methano-5h-inden-5-ylidene bis, 4,4'-octahydro-1h-4,7-methanoindene-5,5-diyldiphenol, p,p'-octahydro-4,7-methano-5h-inden-5-ylidene bisphenol, 4,4'-octahydro-4,7-methano-5h-inden-5-ylidene bisphenol, 4-8-4-hydroxyphenyl tricyclo 5.2.1.0 2 ,? decan-8-yl phenol, dsstox_cid_27869, dsstox_rid_82622, dsstox_gsid_47893
InChI Key WWCSTJWKTAXUGJ-UHFFFAOYSA-N
SMILES C1CC2C(C1)C3CC2CC3(C4=CC=C(C=C4)O)C5=CC=C(C=C5)O
Molecular Weight (g/mol) 320.42
PubChem CID 102724
Formula Weight 320.42
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4,4′-(Octahydro-4,7-methano-5H-inden-5-ylidene) bisphenol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.