Learn More
4,4'-Methylene-bis(2-chloroaniline) 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30325988
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325988 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30325988 Supplier Dr. Ehrenstorfer Supplier No. DREL15087500AL
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4,4'-Methylene-bis(2-chloroaniline)
CAS 101-14-4
Analyte or Component Names 4,4'-Methylene-bis(2-chloroaniline)
Shipping Condition Room Temperature
Molecular Formula C13 H12 Cl2 N2
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzenamine, 4,4′-methylenebis[2-chloro- (9CI, ACI), 4,4′-Methylenebis[2-chlorobenzenamine] (ACI), Aniline, 4,4′-methylenebis[2-chloro- (6CI, 8CI), 2,2′-Dichloro-4,4′-methylenedianiline, 3,3′-Dichloro-4,4′-diaminodiphenylmethane, 4,4′-Methylenebis(2-chloraniline), 4,4′-Methylenebis[2-chloroaniline], 4,4′-Methylenebis[o-chloroaniline], [4-(4-Amino-3-chlorobenzyl)-2-chlorophenyl]amine, Bis(3-chloro-4-aminophenyl)methane, Bis(4-amino-3-chlorophenyl)methane, Bisamine A, Bisamine S, CPP 100, Cuamine K, Cuamine M, Cuamine MT, Curalon M, Curene 442, Diamet Kh, Diamet X, DKh, HR 100, HR 100 (amine), Ihara Cuamine MT, Isocross SM, Kuralon, MBOCA, Methylenebis(3-chloro-4-aminobenzene), Methylenebis(chloraniline), Methylenebis(o-chloroaniline), Millionate M, MOCA, MOCA (curing agent), NSC 52954, Pandex E, Pandex M 3202, Quodorole, SL 4037, Vibracure 133, Vibracure A 133
InChI Formula InChI=1S/C13H12Cl2N2/c14-10-6-8(1-3-12(10)16)5-9-2-4-13(17)11(15)7-9/h1-4,6-7H,5,16-17H2
Solution Type Single Solution
SMILES Nc1ccc(Cc2ccc(N)c(Cl)c2)cc1Cl
IUPAC Name 4-[(4-amino-3-chlorophenyl)methyl]-2-chloroaniline
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 267.15
Quantity 10 mL
Formula Weight 266.0378 g/mol