missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Ethyne-1,2-Diyl)Dibenzoic Acid, TRC

Product Code. 30371821
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30371821 50 mg
30381359 500 mg
30375807 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30371821 Supplier Toronto Research Chemicals Supplier No. TRCE94141850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16819-43-5
Molecular Formula C16H10O4
Molecular Weight (g/mol) 266.25
SMILES OC(=O)c1ccc(cc1)C#Cc2ccc(cc2)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H10O4/c17-15(18)13-7-3-11(4-8-13)1-2-12-5-9-14(10-6-12)16(19)20/h3-10H,(H,17,18)(H,19,20)
Formula Weight 266.0579
Chemical Name or Material 4,4'-(Ethyne-1,2-Diyl)Dibenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.