CAS RN 16819-43-5
CAS RN 16819-43-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,4'-(Ethyne-1,2-Diyl)Dibenzoic Acid, TRC
CAS: 16819-43-5 Molecular Formula: C16H10O4 Molecular Weight (g/mol): 266.25 SMILES: OC(=O)c1ccc(cc1)C#Cc2ccc(cc2)C(=O)O