Learn More
4,4-Dibromo-2,4-dihydro-5-methyl-3H-pyrazol-3-one, TRC
Product Code. 30011473
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011473 100 mg 100mg
30008097 25 mg 25mg
30006514 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011473 Supplier Toronto Research Chemicals Supplier No. TRCD425415100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33549-66-5
Molecular Formula C4 H4 Br2 N2 O
Molecular Weight (g/mol) 255.895
Synonym 3H-Pyrazol-3-one, 4,4-dibromo-2,4-dihydro-5-methyl-, 2-Pyrazolin-5-one, 4,4-dibromo-3-methyl- (6CI,8CI), 4,4-Dibromo-2,4-dihydro-5-methyl-3H-pyrazol-3-one, 4,4-Dibromo-3-methyl-2-pyrazolin-5-one, 4,4-Dibromo-3-methyl-5-pyrazolone, 4,4-Dibromo-3-methyl-δ2-pyrazol-5-one, NSC 522042
IUPAC Name 4,4-dibromo-3-methyl-1H-pyrazol-5-one
SMILES CC1=NNC(=O)C1(Br)Br
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C4H4Br2N2O/c1-2-4(5,6)3(9)8-7-2/h1H3,(H,8,9)
Formula Weight 253.869
Chemical Name or Material 4,4-Dibromo-2,4-dihydro-5-methyl-3H-pyrazol-3-one