Learn More
4,4'-Diaminostilbene, TRC
Specifications
Specifications
| Chemical Name or Material | 4,4'-Diaminostilbene |
| CAS | 621-96-5 |
| Molecular Formula | C14H14N2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C14H14N2/c15-13-7-3-11(4-8-13)1-2-12-5-9-14(16)10-6-12/h1-10H,15-16H2 |
| SMILES | NC1=CC=C(C=C1)C=CC2=CC=C(N)C=C2 |
| IUPAC Name | 4,4'-(ethene-1,2-diyl)dianiline |
| Molecular Weight (g/mol) | 210.27 |
| Formula Weight | 210.12 |