missing translation for 'onlineSavingsMsg'
Learn More

4,4′-Diaminodiphenyl Sulfone, Sigma-Aldrich™

Product Code. 15874591
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874591 250 g 250g
15864591 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15874591 Supplier Sigma Aldrich Supplier No. 8210730250
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-08-0
Grade Research
Molecular Formula [4-(NH2)C6H4]2SO2
Molecular Weight (g/mol) 248.3

Specifications

Melting Point 172°C to 175°C
MDL Number MFCD00007887
Quantity 250 g
Synonym 4,4′-Diaminodiphenyl sulfone, DDS, 4,4′-Sulfonyldianiline, Bis(p-aminophenyl) sulfone, 4,4′-Diaminodiphenyl sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.