Learn More
4,4'-Di-tert-butyl-2,2'-bipyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 4,4'-Di-tert-butyl-2,2'-bipyridine |
| CAS | 72914-19-3 |
| Molecular Formula | C18H24N2 |
| Quantity | 10 g |
| Synonym | 4,4'-Bis(1,1-dimethylethyl)-2,2'-bipyridine, 4,4'-Bis(t-butyl)-2,2'-bipyridine, 4,4'-Bis(tert-butyl)-2,2'-bipyridine, 4,4'-Di-t-butyl-2,2'-bipyridyl, 4,4'-Di-tert-butyl-2,2'-bipyridine, 4,4'-Di-tert-butyl-2,2'-bipyridyl 4,4'-Di-tert-butyl-2,2'-dipyridyl, 4-tert-Butyl-2-(4-tert-butylpyridin-2-yl)pyridine BBBPY |
| InChI Formula | InChI=1S/C18H24N2/c1-17(2,3)13-7-9-19-15(11-13)16-12-14(8-10-20-16)18(4,5)6/h7-12H,1-6H3 |
| SMILES | CC(C)(C)C1=CC(=NC=C1)C2=NC=CC(=C2)C(C)(C)C |
| Molecular Weight (g/mol) | 268.4 |
| Formula Weight | 268.1939 |