Learn More
4,4'-DDT, Fluka™
Product Code. 16222852 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16222852 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16222852 Supplier Honeywell-Fluka Supplier No. R1605100MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Analytical standard, reference material
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4,4′-DDT
Packaging Glass Bottle
CAS 50-29-3
Molecular Formula (ClC6H4)2CHCCl3
Color Colorless
Melting Point 107°C to 110°C
Boiling Point 260°C
Synonym 1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane
Industry Type Environmental Analysis
Molecular Weight (g/mol) 354.49
Quantity 100 mg
Grade Analytical Standard