Learn More
4-(4-Chlorophenyl)but-3-en-2-one, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 4-(4-Chlorophenyl)but-3-en-2-one |
| CAS | 3160-40-5 |
| Molecular Formula | C10H9OCl |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C10H9ClO/c1-8(12)2-3-9-4-6-10(11)7-5-9/h2-7H,1H3/b3-2+ |
| SMILES | CC(=O)\C=C\c1ccc(Cl)cc1 |
| IUPAC Name | (E)-4-(4-chlorophenyl)but-3-en-2-one |
| Molecular Weight (g/mol) | 180.63 |
| Formula Weight | 180.0342 |